siRNA Details
| Gene Symbol: PLA2G2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022819.3 | NP_073730.3 | 4, 5 | 517 | ||
| XM_011541956.1 | XP_011540258.1 | 4, 5 | 521 | |||
| GenBank mRNA | AF306566.1 | - | 3, 4 | 290 | ||
| AK093645.1 | - | 4, 5 | 521 | |||

PLA2G2F
Genomic Map