siRNA Details
| Gene Symbol: PLA2G2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022819.3 | NP_073730.3 | 5 | 602 | ||
| XM_011541956.1 | XP_011540258.1 | 5 | 606 | |||
| GenBank mRNA | AF306566.1 | - | 4 | 375 | ||
| AK093645.1 | - | 5 | 606 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G2F| Gene Symbol: PLA2G2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022819.3 | NP_073730.3 | 5 | 602 | ||
| XM_011541956.1 | XP_011540258.1 | 5 | 606 | |||
| GenBank mRNA | AF306566.1 | - | 4 | 375 | ||
| AK093645.1 | - | 5 | 606 | |||