siRNA Details
| Gene Symbol: MFSD14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033055.2 | NP_149044.2 | 9 | 913 | ||
| XM_005271118.1 | XP_005271175.1 | 10 | 1189 | |||
| XM_011541957.1 | XP_011540259.1 | 8 | 954 | |||
| XM_017002084.1 | XP_016857573.1 | 10 | 1156 | |||
| GenBank mRNA | AF427492.1 | - | 9 | 913 | ||
| AK057172.1 | - | 9 | 1049 | |||
| AK075208.1 | - | 3 | 114 | |||
| AK096669.1 | - | 6 | 564 | |||
| BC064409.1 | - | 6 | 570 | |||
| BC107768.1 | - | 6 | 662 | |||

MFSD14A
Genomic Map