siRNA Details
| Gene Symbol: CSRNP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320559.1 | NP_001307488.1 | 2 | 358 | ||
| NM_001320560.1 | NP_001307489.1 | 2 | 159 | |||
| NM_033027.3 | NP_149016.2 | 2 | 227 | |||
| XM_017007048.1 | XP_016862537.1 | 2 | 3323 | |||
| GenBank mRNA | AB053121.1 | - | 2 | 204 | ||
| AB063301.1 | - | 2 | 119 | |||
| BC038949.1 | - | 2 | 174 | 5547938 | ||
| BG536908.1 | - | 2 | 228 | |||
| BX397882.2 | - | 2 | 159 | |||

CSRNP1
Genomic Map