siRNA Details
| Gene Symbol: MARC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022746.3 | NP_073583.3 | 4, 5 | 1009 | ||
| XM_017002096.1 | XP_016857585.1 | 4, 5 | 589 | |||
| XM_017002097.1 | XP_016857586.1 | 4, 5 | 642 | |||
| GenBank mRNA | AK000035.1 | - | 1, 2 | 31 | ||
| AK026043.1 | - | 4, 5 | 805 | |||
| AK092439.1 | - | 3, 4 | 1480 | |||
| AK225416.1 | - | 4, 5 | 805 | |||
| AK290812.1 | - | 4, 5 | 781 | |||
| BC010619.1 | - | 4, 5 | 777 | 3872779 | ||

MARC1
Genomic Map