siRNA Details
| Gene Symbol: PARP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022750.3 | NP_073587.1 | 3 | 1541 | ||
| NR_130117.1 | 3 | 1541 | ||||
| XM_005250038.3 | XP_005250095.1 | 3 | 1536 | |||
| XM_005250039.4 | XP_005250096.1 | 3 | 1538 | |||
| XM_005250040.4 | XP_005250097.1 | 3 | 1538 | |||
| XM_011516491.2 | XP_011514793.1 | 3 | 1536 | |||
| GenBank mRNA | AL136766.1 | - | 3 | 1163 | ||
| AL137255.1 | - | 3 | 360 | |||
| BC081541.1 | - | 3 | 1541 | 5296560 | ||

PARP12
Genomic Map