siRNA Details
| Gene Symbol: C6orf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022758.5 | NP_073595.2 | 3 | 753 | ||
| NM_024294.3 | NP_077270.1 | 4 | 951 | |||
| XM_005249298.2 | XP_005249355.1 | 4 | 898 | |||
| GenBank mRNA | AK056964.1 | - | 3 | 668 | ||
| AK313410.1 | - | 4 | 857 | |||
| BC002328.1 | - | 4 | 865 | 3504258 | ||
| BC010184.1 | - | 4 | 865 | 3959622 | ||
| BC075810.1 | - | 4 | 873 | 5535440 | ||

C6orf106
Genomic Map