siRNA Details
| Gene Symbol: ARV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022786.1 | NP_073623.1 | 4, 5 | 710 | ||
| XM_017002106.1 | XP_016857595.1 | 5, 6 | 804 | |||
| GenBank mRNA | AF271780.1 | - | 4, 5 | 710 | ||
| AF290878.1 | - | 4, 5 | 710 | |||
| AF321442.1 | - | 4, 5 | 744 | |||
| AK293049.1 | - | 4, 5 | 716 | |||
| AL122047.1 | - | 1, 2 | 109 | |||
| BC016309.1 | - | 4, 5 | 705 | 4047121 | ||

ARV1
Genomic Map