siRNA Details
| Gene Symbol: ISL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145805.2 | NP_665804.1 | 4, 5 | 947 | ||
| XM_017022505.1 | XP_016877994.1 | 4, 5 | 947 | |||
| GenBank mRNA | AB593133.1 | - | 4, 5 | 2469 | ||
| AK001022.1 | - | 4, 5 | 866 | |||
| BC011967.1 | - | 4, 5 | 853 | 3866562 | ||
| BC012136.1 | - | 4, 5 | 865 | 4558235 | ||

ISL2
Genomic Map