siRNA Details
| Gene Symbol: YTHDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022828.3 | NP_073739.3 | 22 | 3232 | ||
| XM_011543581.1 | XP_011541883.1 | 22 | 3243 | |||
| XM_011543582.2 | XP_011541884.1 | 22 | 3217 | |||
| XM_011543586.2 | XP_011541888.1 | 22 | 3217 | |||
| XM_017009730.1 | XP_016865219.1 | 22 | 3243 | |||
| XM_017009731.1 | XP_016865220.1 | 21 | 3018 | |||
| XM_017009732.1 | XP_016865221.1 | 21 | 3018 | |||
| XM_017009733.1 | XP_016865222.1 | 20 | 2846 | |||
| XM_017009734.1 | XP_016865223.1 | 20 | 2846 | |||
| XM_017009735.1 | XP_016865224.1 | 19 | 2665 | |||
| GenBank mRNA | AK025593.1 | - | 6 | 845 | ||
| AK131450.1 | - | 21 | 2925 | |||
| AK225316.1 | - | 6 | 845 | |||
| BC137285.1 | - | 22 | 3182 | 9021282 | ||
| BE295472.1 | - | 2 | 246 | |||

YTHDC2
Genomic Map