siRNA Details
| Gene Symbol: FAM129B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035534.2 | NP_001030611.1 | 10, 11 | 1355 | ||
| NM_022833.3 | NP_073744.2 | 10, 11 | 1567 | |||
| XM_005252135.2 | XP_005252192.2 | 11, 12 | 1414 | |||
| XM_011518925.1 | XP_011517227.1 | 11, 12 | 1586 | |||
| GenBank mRNA | AB210016.1 | - | 10, 11 | 1305 | ||
| AF151783.1 | - | 10, 11 | 1337 | |||
| AF192911.1 | - | 10, 11 | 1558 | |||
| AK023580.1 | - | 2, 3 | 366 | |||
| AL137555.1 | - | 9, 10 | 1175 | |||
| BC001979.1 | - | 3, 4 | 454 | |||

FAM129B
Genomic Map