siRNA Details
| Gene Symbol: MRPL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318767.1 | NP_001305696.1 | 3 | 227 | ||
| NM_001318768.1 | NP_001305697.1 | 3 | 171 | |||
| NM_001318769.1 | NP_001305698.1 | 3 | 160 | |||
| NM_001318770.1 | NP_001305699.1 | 3 | 433 | |||
| NM_001318771.1 | NP_001305700.1 | 2 | 177 | |||
| NM_032111.3 | NP_115487.2 | 3 | 266 | |||
| GenBank mRNA | AK312086.1 | - | 3 | 198 | ||
| BC065005.1 | - | 2 | 122 | 6052682 | ||
| BG545603.1 | - | 3 | 431 | |||
| BM013002.1 | - | 2 | 92 | |||
| BM559405.1 | - | 278 | ||||
| BM910935.1 | - | 229 | ||||
| BU597168.1 | - | 3 | 193 | |||
| BU753526.1 | - | 1 | 500 | |||
| CF541260.1 | - | 1 | 501 | |||
| CK823336.1 | - | 3 | 171 | |||
 Back To Top
 Back To Top
 MRPL14
MRPL14 Genomic Map
Genomic Map Back To Top
 Back To Top