siRNA Details
| Gene Symbol: CENPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022909.3 | NP_075060.1 | 1, 2 | 225 | ||
| GenBank mRNA | AB035124.1 | - | 1, 2 | 138 | ||
| AK123761.1 | - | 1, 2 | 200 | |||
| AK290746.1 | - | 1, 2 | 208 | |||
| BC012024.1 | - | 1, 2 | 201 | 4510607 | ||
| BC015355.1 | - | 1, 2 | 207 | 3451767 | ||
| KU178758.1 | - | 1, 2 | 138 | |||
| KU178759.1 | - | 1, 2 | 138 | |||

CENPH
Genomic Map