siRNA Details
| Gene Symbol: CENPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022909.3 | NP_075060.1 | 8 | 674 | ||
| GenBank mRNA | AB035124.1 | - | 8 | 587 | ||
| AK123761.1 | - | 7 | 592 | |||
| AK290746.1 | - | 8 | 657 | |||
| BC012024.1 | - | 8 | 650 | 4510607 | ||
| BC015355.1 | - | 8 | 656 | 3451767 | ||
| KU178758.1 | - | 8 | 587 | |||
| KU178759.1 | - | 8 | 546 | |||

CENPH
Genomic Map