siRNA Details
| Gene Symbol: VPS33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022916.4 | NP_075067.2 | 4 | 533 | ||
| GenBank mRNA | AF439857.1 | - | 4 | 444 | ||
| AK315545.1 | - | 4 | 471 | |||
| BC016617.1 | - | 4 | 441 | 3855313 | ||
| BI551252.1 | - | 4 | 486 | |||
| BP199163.1 | - | 4 | 533 | |||

VPS33A
Genomic Map