siRNA Details
| Gene Symbol: TMEM135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168724.1 | NP_001162195.1 | 12 | 1269 | ||
| NM_022918.3 | NP_075069.3 | 13 | 1335 | |||
| NR_033149.1 | 12 | 1301 | ||||
| XM_017018140.1 | XP_016873629.1 | 12 | 1200 | |||
| XM_017018141.1 | XP_016873630.1 | 11 | 884 | |||
| XM_017018142.1 | XP_016873631.1 | 11 | 837 | |||
| GenBank mRNA | AK000684.1 | - | 10 | 878 | ||
| AK025757.1 | - | 11 | 883 | |||
| AK225364.1 | - | 11 | 883 | |||
| AK291804.1 | - | 12 | 1179 | |||
| AK296774.1 | - | 9 | 873 | |||
| AK302473.1 | - | 12 | 1105 | |||
| AL834493.1 | - | 4 | 322 | |||
| BC012553.1 | - | 383 | ||||
| BC051462.1 | - | 13 | 1156 | 5278478 | ||
| BX648678.1 | - | 12 | 1255 | |||
| Gene Symbol: CCDC174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BF698254.1 | - | 379 | |||

TMEM135
Genomic Map