siRNA Details
| Gene Symbol: TMEM135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168724.1 | NP_001162195.1 | 8 | 858 | ||
| NM_022918.3 | NP_075069.3 | 9 | 924 | |||
| NR_033149.1 | 8 | 890 | ||||
| XM_017018140.1 | XP_016873629.1 | 8 | 789 | |||
| XM_017018141.1 | XP_016873630.1 | 7 | 473 | |||
| XM_017018142.1 | XP_016873631.1 | 7 | 426 | |||
| GenBank mRNA | AK000684.1 | - | 6 | 468 | ||
| AK025757.1 | - | 7 | 472 | |||
| AK225364.1 | - | 7 | 472 | |||
| AK291804.1 | - | 8 | 768 | |||
| AK296774.1 | - | 5 | 462 | |||
| AK302473.1 | - | 8 | 694 | |||
| BC051462.1 | - | 9 | 745 | 5278478 | ||
| BX648678.1 | - | 8 | 844 | |||

TMEM135
Genomic Map