siRNA Details
| Gene Symbol: UPF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023010.3 | NP_075386.1 | 9 | 1112 | ||
| NM_080632.2 | NP_542199.1 | 10 | 1151 | |||
| XM_017029737.1 | XP_016885226.1 | 10 | 1151 | |||
| XM_017029738.1 | XP_016885227.1 | 10 | 1151 | |||
| XM_017029739.1 | XP_016885228.1 | 9 | 1112 | |||
| XM_017029740.1 | XP_016885229.1 | 10 | 1151 | |||
| GenBank mRNA | AF318576.1 | - | 10 | 1071 | ||
| AK093728.1 | - | 9 | 1050 | |||
| AY013251.1 | - | 9 | 1082 | |||
| BC121017.1 | - | 9 | 1055 | 40117102 | ||

UPF3B
Genomic Map