siRNA Details
| Gene Symbol: FNDC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319152.1 | NP_001306081.1 | 2 | 793 | ||
| NM_001319153.1 | NP_001306082.1 | 2 | 795 | |||
| NM_024059.3 | NP_076964.1 | 2 | 811 | |||
| XM_011529041.2 | XP_011527343.1 | 2 | 1348 | |||
| XM_017028060.1 | XP_016883549.1 | 2 | 947 | |||
| GenBank mRNA | AK125168.1 | - | 1 | 2668 | ||
| BC000912.2 | - | 2 | 811 | 3048411 | ||
| BI828873.1 | - | 2 | 787 | |||
| BM554007.1 | - | 2 | 772 | |||

FNDC11
Genomic Map