siRNA Details
| Gene Symbol: C7orf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303039.1 | NP_001289968.1 | 4 | 810 | ||
| NM_024067.3 | NP_076972.2 | 4 | 867 | |||
| XM_005249845.4 | XP_005249902.1 | 4 | 885 | |||
| XM_011515518.2 | XP_011513820.1 | 4 | 885 | |||
| XM_017012614.1 | XP_016868103.1 | 4 | 1413 | |||
| XM_017012615.1 | XP_016868104.1 | 3 | 409 | |||
| GenBank mRNA | AK056149.1 | - | 4 | 604 | ||
| BC001076.2 | - | 4 | 686 | 2821816 | ||
| BC005121.1 | - | 4 | 686 | 2821816 | ||
| CA454040.1 | - | 1 | 24 | |||

C7orf26
Genomic Map