siRNA Details
| Gene Symbol: FASTKD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024091.3 | NP_076996.2 | 2 | 1094 | ||
| XM_006714498.1 | XP_006714561.1 | 2 | 1005 | |||
| GenBank mRNA | AK026927.1 | - | 2 | 1059 | ||
| BC001295.1 | - | 1 | 586 | |||
| BC113563.1 | - | 1 | 1009 | 8322615 | ||
| BC143517.1 | - | 1 | 1009 | |||
| BC143518.1 | - | 2 | 446 | |||
| BP317791.1 | - | 1 | 34 | |||

FASTKD3
Genomic Map