siRNA Details
| Gene Symbol: CCDC86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024098.3 | NP_077003.1 | 3, 4 | 1126 | ||
| GenBank mRNA | AK025974.1 | - | 3, 4 | 1012 | ||
| AK123775.1 | - | 5, 6 | 799 | |||
| AK302329.1 | - | 4, 5 | 916 | |||
| BC001378.1 | - | 3, 4 | 977 | 3051044 | ||
| BE795888.1 | - | 2, 3 | 97 | |||
| DQ501251.1 | - | 3, 4 | 957 | |||

CCDC86
Genomic Map