siRNA Details
| Gene Symbol: GNPTAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024312.4 | NP_077288.2 | 13 | 1846 | ||
| XM_006719593.2 | XP_006719656.1 | 13 | 1841 | |||
| XM_011538731.2 | XP_011537033.1 | 13 | 1842 | |||
| XM_017019961.1 | XP_016875450.1 | 13 | 1743 | |||
| XM_017019962.1 | XP_016875451.1 | 13 | 1948 | |||
| GenBank mRNA | AB033034.2 | - | 6 | 737 | ||
| AK056137.1 | - | 13 | 1717 | |||
| AM085438.1 | - | 13 | 1833 | |||
| AY687932.1 | - | 13 | 1818 | |||
| BC131787.1 | - | 13 | 1774 | 40126580 | ||

GNPTAB
Genomic Map