siRNA Details
| Gene Symbol: GNPTAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024312.4 | NP_077288.2 | 5 | 652 | ||
| XM_006719593.2 | XP_006719656.1 | 5 | 647 | |||
| XM_011538731.2 | XP_011537033.1 | 5 | 648 | |||
| XM_017019961.1 | XP_016875450.1 | 5 | 549 | |||
| XM_017019962.1 | XP_016875451.1 | 5 | 631 | |||
| GenBank mRNA | AK056137.1 | - | 5 | 523 | ||
| AL832172.1 | - | 1 | 94 | |||
| AM085438.1 | - | 5 | 639 | |||
| AY687932.1 | - | 5 | 624 | |||
| BC071687.1 | - | 5 | 600 | |||
| BC131787.1 | - | 5 | 580 | 40126580 | ||

GNPTAB
Genomic Map