siRNA Details
| Gene Symbol: GNPTAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024312.4 | NP_077288.2 | 7 | 896 | ||
| XM_006719593.2 | XP_006719656.1 | 7 | 891 | |||
| XM_011538731.2 | XP_011537033.1 | 7 | 892 | |||
| XM_017019961.1 | XP_016875450.1 | 7 | 793 | |||
| XM_017019962.1 | XP_016875451.1 | 7 | 875 | |||
| GenBank mRNA | AK056137.1 | - | 7 | 767 | ||
| AM085438.1 | - | 7 | 883 | |||
| AY687932.1 | - | 7 | 868 | |||
| BC071687.1 | - | 7 | 844 | |||
| BC131787.1 | - | 7 | 824 | 40126580 | ||

GNPTAB
Genomic Map