siRNA Details
| Gene Symbol: RBM42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319113.1 | NP_001306042.1 | 8, 9 | 1375 | ||
| NM_024321.4 | NP_077297.2 | 9, 10 | 1462 | |||
| XM_005259257.2 | XP_005259314.1 | 9, 10 | 1436 | |||
| GenBank mRNA | BC002868.2 | - | 8, 9 | 1314 | 3941786 | |
| BC004204.2 | - | 9, 10 | 1400 | 3532802 | ||
| BC031682.1 | - | 8, 9 | 1339 | 5163682 | ||
| BU732481.1 | - | 1, 2 | 266 | |||

RBM42
Genomic Map