siRNA Details
| Gene Symbol: ZNF557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044387.1 | NP_001037852.1 | 6 | 771 | ||
| NM_001044388.1 | NP_001037853.1 | 6 | 762 | |||
| NM_024341.2 | NP_077317.2 | 6 | 783 | |||
| GenBank mRNA | AK095524.1 | - | 6 | 516 | ||
| AK297283.1 | - | 6 | 540 | |||
| AK315405.1 | - | 6 | 800 | |||
| BC003020.1 | - | 6 | 533 | 2822966 | ||
| BC031208.1 | - | 6 | 540 | |||
| BC058040.1 | - | 6 | 512 | |||

ZNF557
Genomic Map