siRNA Details
| Gene Symbol: ZXDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040653.3 | NP_001035743.1 | 6 | 3997 | ||
| NM_025112.4 | NP_079388.3 | 7 | 2200 | |||
| XM_005247757.3 | XP_005247814.1 | 7 | 2211 | |||
| XM_011513119.2 | XP_011511421.1 | 6 | 2048 | |||
| GenBank mRNA | AF131763.1 | - | 1 | 1330 | ||
| AK023923.1 | - | 4 | 902 | |||
| AY277596.1 | - | 3 | 762 | |||
| AY936556.1 | - | 7 | 2160 | |||
| BC002940.2 | - | 6 | 1138 | |||
| BC012729.2 | - | 6 | 1138 | |||
| DQ079590.1 | - | 7 | 2200 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_173002, NM_030260, AK047538, AK169517, AK044529, and others.

ZXDC
Genomic Map