siRNA Details
| Gene Symbol: B3GNT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030765.2 | NP_110392.1 | 3 | 981 | ||
| XM_011538732.1 | XP_011537034.1 | 2 | 818 | |||
| GenBank mRNA | AB049586.1 | - | 2 | 851 | ||
| AJ130848.1 | - | 1 | 688 | |||
| AJ278962.1 | - | 1 | 688 | |||
| AK095746.1 | - | 2 | 818 | |||
| AL834452.1 | - | 2 | 795 | |||
| AY358940.1 | - | 3 | 1119 | |||
| BC031399.1 | - | 3 | 948 | 5197310 | ||

B3GNT4
Genomic Map