siRNA Details
| Gene Symbol: GLB1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286423.1 | NP_001273352.1 | 14 | 1529 | ||
| NM_001286427.1 | NP_001273356.1 | 11 | 1046 | |||
| NM_024506.4 | NP_078782.3 | 14 | 1517 | |||
| XM_011511813.2 | XP_011510115.1 | 12 | 1106 | |||
| XM_017004895.1 | XP_016860384.1 | 12 | 1213 | |||
| GenBank mRNA | AK055098.1 | - | 9 | 704 | ||
| AK074393.1 | - | 8 | 697 | |||
| AK299404.1 | - | 13 | 1278 | |||
| AK300215.1 | - | 11 | 1029 | |||
| AY358639.1 | - | 14 | 1497 | |||

GLB1L
Genomic Map