siRNA Details
| Gene Symbol: LRRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024512.4 | NP_078788.2 | 5, 6 | 1016 | ||
| XM_011534110.2 | XP_011532412.1 | 5, 6 | 805 | |||
| XM_017007177.1 | XP_016862666.1 | 5, 6 | 1016 | |||
| GenBank mRNA | AJ308569.2 | - | 5, 6 | 790 | ||
| AK057808.1 | - | 5, 6 | 858 | |||
| AK223440.1 | - | 5, 6 | 858 | |||
| AK314658.1 | - | 5, 6 | 779 | |||
| AK315636.1 | - | 5, 6 | 799 | |||
| BC029118.1 | - | 5, 6 | 788 | 5183639 | ||

LRRC2
Genomic Map