siRNA Details
| Gene Symbol: GCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024523.5 | NP_078799.2 | 2 | 2315 | ||
| GenBank mRNA | AF525417.1 | - | 2 | 2090 | ||
| AK025688.1 | - | 2 | 974 | |||
| AK092878.1 | - | 2 | 1771 | |||
| AK315472.1 | - | 2 | 2166 | |||
| AL832833.1 | - | 2 | 977 | |||
| BC008902.2 | - | 2 | 1097 | |||
| BC014100.2 | - | 2 | 2315 | 2988591 | ||
| BC078665.1 | - | 2 | 2267 | 5923117 | ||

GCC1
Genomic Map