siRNA Details
| Gene Symbol: C22orf46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142964.1 | NP_001136436.1 | 2 | 3826 | ||
| XM_011530373.2 | XP_011528675.1 | 3 | 3734 | |||
| XM_017028939.1 | XP_016884428.1 | 4 | 3865 | |||
| GenBank mRNA | AK027237.1 | - | 1 | 739 | ||
| AK131457.1 | - | 4 | 3660 | |||
| BC007210.1 | - | 1 | 821 | 3344580 | ||
| BC035403.1 | - | 2 | 3826 | |||

C22orf46
Genomic Map