siRNA Details
| Gene Symbol: ARSJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024590.3 | NP_078866.3 | 2 | 2402 | ||
| XM_005263212.4 | XP_005263269.1 | 2 | 2360 | |||
| XM_011532238.2 | XP_011530540.1 | 3 | 1739 | |||
| XM_011532239.2 | XP_011530541.1 | 4 | 1277 | |||
| XM_017008592.1 | XP_016864081.1 | 3 | 2442 | |||
| XM_017008593.1 | XP_016864082.1 | 6 | 2997 | |||
| XM_017008594.1 | XP_016864083.1 | 5 | 2564 | |||
| XM_017008595.1 | XP_016864084.1 | 6 | 2677 | |||
| XM_017008596.1 | XP_016864085.1 | 6 | 2657 | |||
| XM_017008597.1 | XP_016864086.1 | 6 | 2688 | |||
| XM_017008598.1 | XP_016864087.1 | 4 | 2514 | |||
| XM_017008599.1 | XP_016864088.1 | 4 | 2523 | |||
| XM_017008600.1 | XP_016864089.1 | 3 | 1227 | |||
| GenBank mRNA | AK027201.1 | - | 1 | 427 | ||
| AK307532.1 | - | 4 | 2044 | |||
| AM049401.1 | - | 2 | 1676 | |||
| AY358647.1 | - | 2 | 2403 | |||
| AY875938.1 | - | 2 | 1514 | |||
| BC032010.1 | - | 1 | 479 | |||
| BC033156.1 | - | 2 | 1306 | |||
| BC089445.1 | - | 2 | 1772 | |||
| BC132879.1 | - | 2 | 1602 | 40146901 | ||
| BC132881.1 | - | 2 | 1602 | 40146903 | ||
| BC144265.1 | - | 2 | 1602 | 9052785 | ||
| BC151213.1 | - | 3 | 2618 | |||

ARSJ
Genomic Map