siRNA Details
| Gene Symbol: HSPBAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024610.5 | NP_078886.2 | 6 | 904 | ||
| XM_011513130.2 | XP_011511432.1 | 7 | 822 | |||
| XM_011513132.2 | XP_011511434.2 | 6 | 663 | |||
| XM_017007179.1 | XP_016862668.1 | 5 | 728 | |||
| XM_017007180.1 | XP_016862669.1 | 2 | 366 | |||
| GenBank mRNA | AF400663.1 | - | 6 | 871 | ||
| AK289410.1 | - | 6 | 904 | |||
| BC011897.2 | - | 6 | 888 | 3949150 | ||

HSPBAP1
Genomic Map