siRNA Details
| Gene Symbol: CENPU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024629.3 | NP_078905.2 | 3 | 193 | ||
| NR_104593.1 | 3 | 193 | ||||
| XM_005263218.4 | XP_005263275.2 | 3 | 228 | |||
| GenBank mRNA | AF469667.1 | - | 3 | 196 | ||
| AF516710.1 | - | 4 | 610 | |||
| AK292297.1 | - | 3 | 139 | |||
| BC131556.1 | - | 3 | 193 | 40108113 | ||
| DQ907910.1 | - | 3 | 123 | |||
| DQ907911.1 | - | 3 | 123 | |||

CENPU
Genomic Map