siRNA Details
| Gene Symbol: ZYG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024646.2 | NP_078922.1 | 6 | 1510 | ||
| XM_006710898.3 | XP_006710961.1 | 6 | 1397 | |||
| XM_017002336.1 | XP_016857825.1 | 6 | 1515 | |||
| GenBank mRNA | AB051517.1 | - | 6 | 1460 | ||
| AK056645.1 | - | 4 | 454 | |||
| AK095643.1 | - | 5 | 1150 | |||
| AK296460.1 | - | 6 | 1515 | |||
| BC029832.1 | - | 4 | 524 | |||
| BX647643.1 | - | 4 | 683 | |||
| DQ646393.1 | - | 6 | 1529 | |||

ZYG11B
Genomic Map