siRNA Details
Gene Symbol: MORC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001085354.2 | NP_001078823.1 | 5 | 844 | ||
NM_024657.4 | NP_078933.3 | 5 | 844 | |||
XM_005262190.3 | XP_005262247.1 | 4 | 734 | |||
XM_011531027.2 | XP_011529329.1 | 2 | 132 | |||
XM_017029844.1 | XP_016885333.1 | 4 | 734 | |||
GenBank mRNA | AK056235.1 | - | 1 | 56 | ||
AK300305.1 | - | 4 | 419 | |||
AY927537.1 | - | LIT3543 AY927537 |
3 | 379 | ||
EF125209.1 | - | 5 | 569 |
