siRNA Details
Gene Symbol: ZNF768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024671.3 | NP_078947.3 | 1, 2 | 267 | ||
GenBank mRNA | AK027089.1 | - | 1, 2 | 39 | ||
AK315449.1 | - | 1, 2 | 63 | |||
BC013760.1 | - | 1, 2 | 267 | 3856266 | ||
BC092403.1 | - | 1, 2 | 179 | 6201079 |
