siRNA Details
| Gene Symbol: ASB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024701.3 | NP_078977.2 | 6 | 863 | ||
| NR_024581.1 | 5 | 712 | ||||
| NR_037164.1 | 7 | 944 | ||||
| GenBank mRNA | AF403031.1 | - | 6 | 845 | ||
| AK023196.1 | - | 6 | 846 | |||
| AK091935.1 | - | 5 | 712 | |||
| AK292071.1 | - | 6 | 849 | |||
| BC012056.1 | - | 7 | 921 | 4552643 | ||
| CR457302.1 | - | 6 | 819 | |||

ASB13
Genomic Map