siRNA Details
| Gene Symbol: RERGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286201.1 | NP_001273130.1 | 5 | 425 | ||
| NM_024730.3 | NP_079006.1 | 6 | 582 | |||
| NR_104413.1 | 5 | 322 | ||||
| GenBank mRNA | AA220227.1 | - | 5 | 269 | ||
| AK026308.1 | - | 6 | 508 | |||
| BC042888.1 | - | 6 | 569 | 5198817 | ||
| BX422875.2 | - | 5 | 373 | |||
| DT220095.1 | - | 6 | 582 | |||

RERGL
Genomic Map