siRNA Details
| Gene Symbol: C12orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024738.2 | NP_079014.1 | 4 | 672 | ||
| XM_005253937.3 | XP_005253994.1 | 3 | 509 | |||
| GenBank mRNA | AK025068.1 | - | 4 | 640 | ||
| AK222983.1 | - | 4 | 652 | |||
| AK298857.1 | - | 3 | 514 | |||
| AK309652.1 | - | 3 | 346 | |||
| BC019843.1 | - | 4 | 532 | 5112874 | ||

C12orf49
Genomic Map