siRNA Details
| Gene Symbol: ZNF408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184751.1 | NP_001171680.1 | 2 | 146 | ||
| NM_024741.2 | NP_079017.1 | 2 | 431 | |||
| GenBank mRNA | AF346626.1 | - | 2 | 381 | ||
| AK022889.1 | - | 2 | 380 | |||
| AK302180.1 | - | 2 | 146 | |||
| BC013355.2 | - | 2 | 153 | 4040144 | ||
| BC015708.2 | - | 2 | 160 | 3911552 | ||
| DA015297.1 | - | 2 | 432 | |||

ZNF408
Genomic Map