siRNA Details
| Gene Symbol: ZNF552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024762.3 | NP_079038.2 | 3 | 941 | ||
| XM_005259267.3 | XP_005259324.1 | 3 | 1119 | |||
| GenBank mRNA | AK025256.1 | - | 1 | 245 | ||
| AK097041.1 | - | 3 | 941 | |||
| AK225219.1 | - | 1 | 245 | |||
| AK293445.1 | - | 3 | 1120 | |||
| BC017460.1 | - | 1 | 379 | |||
| BC046237.1 | - | 1 | 379 | |||
| BC062982.1 | - | 1 | 379 | |||
| BC082767.1 | - | 1 | 379 | |||

ZNF552
Genomic Map