siRNA Details
| Gene Symbol: CATSPERB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024764.3 | NP_079040.2 | 19 | 2105 | ||
| GenBank mRNA | AK024360.1 | - | 5 | 633 | ||
| AK126034.1 | - | 19 | 2089 | |||
| BC126214.1 | - | 19 | 2059 | 8991930 | ||
| BC126216.1 | - | 19 | 2059 | 8991932 | ||

CATSPERB
Genomic Map