siRNA Details
| Gene Symbol: NHEJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024782.2 | NP_079058.1 | 3 | 502 | ||
| GenBank mRNA | AJ972687.1 | - | 3 | 446 | ||
| AK022672.1 | - | 3 | 438 | |||
| BC012732.2 | - | 3 | 423 | 4026495 | ||
| BC030986.1 | - | 3 | 455 | 4076851 | ||
| CR457291.1 | - | 2 | 356 | |||
| DB124934.1 | - | 3 | 342 | |||

NHEJ1
Genomic Map