siRNA Details
| Gene Symbol: EPHX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142886.1 | NP_001136358.1 | 7 | 1013 | ||
| NM_024794.2 | NP_079070.1 | 6 | 1049 | |||
| GenBank mRNA | AK026061.1 | - | 6 | 1049 | ||
| BC115002.1 | - | 7 | 923 | 7482856 | ||
| BC132958.1 | - | 6 | 1013 | 40146980 | ||
| BC132960.1 | - | 6 | 1013 | 40146982 | ||

EPHX3
Genomic Map