siRNA Details
| Gene Symbol: TUBAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171864.1 | NP_001165335.1 | 2 | 75 | ||
| NM_024803.2 | NP_079079.1 | 2 | 195 | |||
| GenBank mRNA | AK025318.1 | - | 2 | 197 | ||
| AK296616.1 | - | 2 | 73 | |||
| BC098247.1 | - | 2 | 68 | 40005930 | ||
| BC099716.1 | - | 2 | 188 | 40005934 | ||
| BC105634.1 | - | 2 | 188 | 40005931 | ||
| DA890426.1 | - | 2 | 200 | |||

TUBAL3
Genomic Map