siRNA Details
Gene Symbol: RABEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024816.2 | NP_079092.2 | 6, 7 | 1026 | ||
GenBank mRNA | AK026935.1 | - | 6, 7 | 1026 | ||
AK092299.1 | - | 6, 7 | 1567 | |||
AK295229.1 | - | 5, 6 | 798 | |||
BC027584.1 | - | 6, 7 | 1504 | 4827815 | ||
BC046118.1 | - | 6, 7 | 1020 |
