siRNA Details
| Gene Symbol: RABEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024816.2 | NP_079092.2 | 8 | 1177 | ||
| GenBank mRNA | AK026935.1 | - | 8 | 1177 | ||
| AK092299.1 | - | 8 | 1718 | |||
| AK295229.1 | - | 7 | 949 | |||
| BC046118.1 | - | 8 | 1171 | |||
| BC058900.1 | - | 7 | 1084 | 5415624 | ||

RABEP2
Genomic Map